Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
4-Fluoro-3-nitrobenzenesulphonyl chloride
4-Fluoro-3-nitrobenzenesulphonyl chloride 99%
4-Fluoro-3-nitrobenzenesulphonyl chloride 99%
:
Apollo Scientific
4-Fluoro-3-nitrobenzenesulphonyl chloride 99%
:
Apollo Scientific
:
6668-56-0
:
:
C₆H₃ClFNO₄S
:
MFCD06342770
:
239.6100 g/mol
:
124…126 °C/0.3mmHg
:
Ambient
:
1760
:
8,III
:
GHS05