Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4-fluorophenol
2-Amino-4-fluorophenol 98%
2-Amino-4-fluorophenol 98%
Irritant
2-Amino-4-fluorophenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-4-fluorophenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
399-97-3
:
Formula:
C₆H₆FNO
MDL Number:
MFCD00077451
:
127.12 g/mol
Melting Pt:
138…141 °C
:
GHS07
Storage Temperature:
Ambient