Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-chloro-4-fluoroanisole
5-Bromo-2-chloro-4-fluoroanisole
5-Bromo-2-chloro-4-fluoroanisole
:
Apollo Scientific
5-Bromo-2-chloro-4-fluoroanisole
:
Apollo Scientific
:
146447-18-9
:
:
C₇H₅BrClFO
:
MFCD07784265
:
239.47 g/mol
:
GHS07
:
Ambient