Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoroisonicotinic acid
2,3-Difluoroisonicotinic acid 97%
2,3-Difluoroisonicotinic acid 97%
Irritant
2,3-Difluoroisonicotinic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Difluoroisonicotinic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
851386-31-7
:
Formula:
C₆H₃F₂NO₂
MDL Number:
MFCD09031174
:
159.09 g/mol
Melting Pt:
163…168 °C
:
GHS07
Storage Temperature:
Refrigerator