

Perfluoro(4-methyl-2-pentenoic acid) 97%
Catalog # APOSPC4808S-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:276.06 g/mol
Boiling Pt:173…174 °C
Density:1.6 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
:GHS07
CAS Number:103229-89-6
:239795-58-5
Formula:C₆HF₉O₂
MDL Number:MFCD00153286