2-(2-Fluorophenoxy)-1-ethanamine
Catalog # APOSPC430517-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:155.17 g/mol
Storage Temperature:Ambient
CAS Number:120351-90-8
Formula:C₈H₁₀FNO
MDL Number:MFCD00235182