Ethyl-3-amino-4,4,4-trifluorobutyrate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:170804-18-9
:
Formula:C₆H₁₀F₃NO₂
MDL Number:MFCD01075754
:185.15 g/mol
Boiling Pt:70…71 °C/4mmHg
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02