4-Chloro-6-fluoroquinazoline
Catalog # APOSPC400552-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.59 g/mol
Storage Temperature:Ambient
CAS Number:16499-61-9
Formula:C₈H₄ClFN₂
MDL Number:MFCD07776652