

4-Fluoro-3-nitrobenzotrifluoride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367-86-2
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD00007059
:209.1 g/mol
Boiling Pt:91…92 °C/15mmHg
Density:1.494 g/cm³ (25 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
EINECS:206-702-5
UN:1993
ADR:3,III
:GHS02
:GHS07