Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,3-difluorobenzoic acid
4-Chloro-2,3-difluorobenzoic acid 97%
4-Chloro-2,3-difluorobenzoic acid 97%
4-Chloro-2,3-difluorobenzoic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloro-2,3-difluorobenzoic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
150444-94-3
:
Formula:
C₇H₃ClF₂O₂
MDL Number:
MFCD07368122
:
192.546 g/mol
Storage Temperature:
Ambient