Order Entry
Norway
ContactUsLinkComponent
 
2-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302925-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302925-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.59 g/mol
Storage Temperature:Ambient
CAS Number:1017778-66-3
Formula:C₉H₆ClF₃O₂
MDL Number:MFCD09832312