Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-2,3-difluorophenylacetic acid
6-Chloro-2,3-difluorophenylacetic acid 98%
6-Chloro-2,3-difluorophenylacetic acid 98%
6-Chloro-2,3-difluorophenylacetic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloro-2,3-difluorophenylacetic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
887585-13-9
:
Formula:
C₈H₅ClF₂O₂
MDL Number:
MFCD07368098
:
206.57 g/mol
Storage Temperature:
Ambient