4-Methoxy-3-(trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:694514-21-1
:
Formula:C₈H₆ClF₃O₃S
MDL Number:MFCD07774167
Melting Pt:52…55 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:274.64 g/mol