
2-Fluoro-5-formylbenzoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:550363-85-4
:
Formula:C₈H₅FO₃
MDL Number:MFCD04972875
:168.13 g/mol
Melting Pt:174…179 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07