1-Bromo-4-(difluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5905-69-1
:
Formula:C₇H₅BrF₂O
MDL Number:MFCD00221465
:223.02 g/mol
Boiling Pt:96…98 °C/15mmHg
Density:1.631 g/cm³ (25 °C)
Flash Pt:34 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07