kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
3,5-Difluorobenzoic acid
3,5-Difluorobenzoic acid 98%
3,5-Difluorobenzoic acid 98%
:
Apollo Scientific
3,5-Difluorobenzoic acid 98%
:
Apollo Scientific
:
455-40-3
:
:
C₇H₄F₂O₂
:
MFCD00010323
:
158.1 g/mol
:
119…123 °C
:
610-261-8
:
GHS07
:
Ambient