
2',4'-Difluoroacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:364-83-0
:
Formula:C₈H₆F₂O
MDL Number:MFCD00151261
:156.13 g/mol
Boiling Pt:80…81 °C/25mmHg
Density:1.232 g/cm³ (25 °C)
Flash Pt:66 °C
EINECS:206-667-6
:GHS07
Storage Temperature:Ambient