Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,8-Dichloro-2-(trifluoromethyl)quinoline
4,8-Dichloro-2-(trifluoromethyl)quinoline 97%
4,8-Dichloro-2-(trifluoromethyl)quinoline 97%
APOSPC2556X-5G
:
Apollo Scientific
:
4,8-Dichloro-2-(trifluoromethyl)quinoline 97%
APOSPC2556X-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
266.0467 g/mol
:
60…63 °C
:
GHS07
:
Ambient
:
18706-35-9
:
C₁₀H₄Cl₂F₃N
:
MFCD00276590