
3-Chloro-2,4,5,6-tetrafluoropyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1735-84-8
:
Formula:C₅ClF₄N
MDL Number:MFCD00010624
:185.51 g/mol
Boiling Pt:118…121 °C
Density:1.609 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient