kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Potassium (3-pyridyl)trifluoroborate
Potassium(3-pyridyl)trifluoroborate
Potassium(3-pyridyl)trifluoroborate
Potassium(3-pyridyl)trifluoroborate
Supplier:
Apollo Scientific
Certificates
Expand image
Potassium(3-pyridyl)trifluoroborate
Supplier:
Apollo Scientific
About this item
CAS Number:
561328-69-6
:
Formula:
C₅H₄BF₃KN
MDL Number:
MFCD09038545
:
185.0000 g/mol
Storage Temperature:
Ambient