Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoroisonicotinic acid
3,5-Difluoroisonicotinic acid
3,5-Difluoroisonicotinic acid
Irritant
3,5-Difluoroisonicotinic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Difluoroisonicotinic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
903522-29-2
:
Formula:
C₆H₃F₂NO₂
MDL Number:
MFCD04972829
:
159.0900 g/mol
:
GHS07
Storage Temperature:
Ambient