ω-Chloro-ω,ω-difluoroacetophenone 97%
Glass bottle, 1 g
Catalog # APOSPC1753-1G
Supplier: Apollo Scientific
ω-Chloro-ω,ω-difluoroacetophenone 97%
Catalog # APOSPC1753-1G
Supplier: Apollo Scientific
CAS Number: 384-67-8
Jump to:
Product Details & Documents
Synonyms: 2-Chloro-2,2-difluoro-1-phenylethanone, α-Chloro-α,α-difluoroacetophenone, 2-Chloro-2,2-difluoroacetophenone
- CAS Number:384-67-8
- Formula:(C₆H₅)C(O)CClF₂
- MDL Number:MFCD00042123
- Molecular Weight:190.58 g/mol
- Boiling Pt:95…97 °C/37mmHg
- Density:1.293 g/cm³ (25 °C)
- Flash Pt:72 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



