kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Potassium (3,5-difluorophenyl)trifluoroborate
Potassium(3,5-difluorophenyl)trifluoroborate 96%
Potassium(3,5-difluorophenyl)trifluoroborate 96%
APOSPC1724-5G
:
Apollo Scientific
:
Potassium(3,5-difluorophenyl)trifluoroborate 96%
APOSPC1724-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
219.99 g/mol
:
Ambient
:
267006-26-8
:
C₆H₃BF₅K
:
MFCD06659939