Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,5-Trifluorophenylboronic acid
2,3,5-Trifluorophenylboronic acid 95%
2,3,5-Trifluorophenylboronic acid 95%
APOSPC1511-5G
:
Apollo Scientific
:
2,3,5-Trifluorophenylboronic acid 95%
APOSPC1511-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
175.9 g/mol
:
221 °C (dec.)
:
GHS07
:
Ambient
:
247564-73-4
:
C₆H₄BF₃O₂
:
MFCD01863166