Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-2,5-dichloro-3-fluorobenzene
1-Bromo-2,5-dichloro-3-fluorobenzene 98%
1-Bromo-2,5-dichloro-3-fluorobenzene 98%
Irritant
1-Bromo-2,5-dichloro-3-fluorobenzene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Bromo-2,5-dichloro-3-fluorobenzene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
202865-57-4
:
Formula:
C₆H₂BrCl₂F
MDL Number:
MFCD00142585
:
243.89 g/mol
:
GHS07
Storage Temperature:
Ambient