1-Bromo-4-chloro-3-fluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60811-18-9
:
Formula:C₆H₃BrClF
MDL Number:MFCD00042519
:209.44 g/mol
Boiling Pt:62…64 °C/9mmHg
Density:1.71 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient