
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16627-71-7
:
Formula:C₇H₄F₁₂O
MDL Number:MFCD00155904
:332.09 g/mol
Boiling Pt:133 °C
:GHS07
Storage Temperature:Ambient