
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:88738-78-7
:
Formula:C₇H₉F₆O₅P
MDL Number:MFCD00009901
:318.11 g/mol
Density:1.504 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together