(R)-(+)-4-Fluoro-α-methylbenzyl alcohol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:101219-68-5
:
Formula:C₈H₉FO
MDL Number:MFCD03092997
Boiling Pt:90…92 °C/6,75mmHg
Density:1,109 g/cm³ (25 °C)
Flash Pt:91 °C
:140.16 g/mol
:GHS07
Storage Temperature:Romtemperatur