Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-fluorothiophenol
2-Bromo-4-fluorothiophenol
2-Bromo-4-fluorothiophenol
Irritant
2-Bromo-4-fluorothiophenol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-4-fluorothiophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
773853-92-2
:
Formula:
C₆H₄BrFS
MDL Number:
MFCD09475472
:
GHS07
Storage Temperature:
Refrigerator