Order Entry
Norway
ContactUsLinkComponent
4-Isopropyl-1,3-thiazol-2-amine hydrobromide 98%
4-Isopropyl-1,3-thiazol-2-amine hydrobromide 98%
Catalog # APOSOR965018-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Isopropyl-1,3-thiazol-2-amine hydrobromide 98%
Catalog # APOSOR965018-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :223.14 g/mol
Storage Temperature:Ambient
CAS Number:79932-20-0
Formula:C₆H₁₁BrN₂S
MDL Number:MFCD21605754