(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:914349-47-6
:
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD05864818
:188.2300 g/mol
Storage Temperature:Ambient
Frequently Bought Together