Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Ethylphenol
3-Ethylphenol 95%
3-Ethylphenol 95%
3-Ethylphenol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Ethylphenol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
620-17-7
:
Formula:
C₈H₁₀O
MDL Number:
MFCD00002311
:
122.17 g/mol
Storage Temperature:
Ambient