Order Entry
Norway
ContactUsLinkComponent
Ethyl-4-formylbenzoate 95%
Ethyl-4-formylbenzoate 95%
Catalog # APOSOR938434-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-4-formylbenzoate 95%
Catalog # APOSOR938434-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.19 g/mol
Storage Temperature:Ambient
CAS Number:6287-86-1
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00460463