Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2305-13-7
:
Formula:
C₁₀H₁₄O₃
MDL Number:
MFCD00016571
:
182.22 g/mol
Storage Temperature:
Ambient