Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5-(chloromethyl)pyrimidine
2-Chloro-5-(chloromethyl)pyrimidine 95%
2-Chloro-5-(chloromethyl)pyrimidine 95%
2-Chloro-5-(chloromethyl)pyrimidine 95%
Leverandør:
Apollo Scientific
Sertifikat
Expand image
2-Chloro-5-(chloromethyl)pyrimidine 95%
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
148406-13-7
:
Formel:
C₅H₄Cl₂N₂
MDL-nummer:
MFCD10697062
:
163.01 g/mol
Lagringstemperatur:
Ambient