Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-9H-purine
2-Chloro-9H-purine 96%
2-Chloro-9H-purine 96%
2-Chloro-9H-purine 96%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-9H-purine 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
1681-15-8
:
Formula:
C₅H₃ClN₄
MDL Number:
MFCD00137826
:
154.5600 g/mol
Storage Temperature:
Ambient