Order Entry
Norway
ContactUsLinkComponent
1-Methylcyclohexanol 95%
1-Methylcyclohexanol 95%
Catalog # APOSOR925032-100G
Supplier:  Apollo Scientific
CAS Number:  
1-Methylcyclohexanol 95%
Catalog # APOSOR925032-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :114.19 g/mol
Boiling Pt:268 °C/752mmHg
Melting Pt:26 °C
Density:0.919 g/cm³ (25 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
CAS Number:590-67-0
Formula:C₇H₁₄O
MDL Number:MFCD00003857