Order Entry
Norway
ContactUsLinkComponent
L(-)-Menthol 95%
L(-)-Menthol 95%
Katalog # APOSOR921562-500G
Supplier:  Apollo Scientific
CAS Number:  
L(-)-Menthol 95%
Katalog # APOSOR921562-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :156.27 g/mol
Storage Temperature:Ambient
CAS Number:2216-51-5
Formula:C₁₀H₂₀O
MDL Number:MFCD00062979