Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Norvaline ethyl ester hydrochloride
L-Norvaline ethyl ester hydrochloride 95%
L-Norvaline ethyl ester hydrochloride 95%
L-Norvaline ethyl ester hydrochloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
L-Norvaline ethyl ester hydrochloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
40918-51-2
:
Formula:
C₇H₁₅NO₂
MDL Number:
MFCD08458628
:
181.66 g/mol
Storage Temperature:
Ambient