1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98%
Catalog # APOSOR911373-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:277.316 g/mol
Melting Pt:273…278 °C
Storage Temperature:Refrigerator
CAS Number:174543-74-9
Formula:C₁₅H₁₉NO₄
MDL Number:MFCD06795953