Methyl-4-pentenoate 95%
Katalog # APOSOR908012-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.14 g/mol
Density:0.922 g/cm³ (25 °C)
Flash Pt:29 °C
Storage Temperature:Ambient
UN:3272
ADR:3,III
CAS Number:818-57-5
Formula:C₆H₁₀O₂
MDL Number:MFCD03990590