kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Methyl 6-iodonicotinate
Methyl-6-iodonicotinate 96%
Methyl-6-iodonicotinate 96%
Methyl-6-iodonicotinate 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-6-iodonicotinate 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
173157-33-0
:
Formula:
C₇H₆INO₂
MDL Number:
MFCD12546492
:
263.03 g/mol
:
GHS07
Storage Temperature:
Refrigerator