Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dichloro-2-(methylsulphonyl)benzene
1,4-Dichloro-2-(methylsulphonyl)benzene 95+%
1,4-Dichloro-2-(methylsulphonyl)benzene 95+%
:
Apollo Scientific
1,4-Dichloro-2-(methylsulphonyl)benzene 95+%
:
Apollo Scientific
:
66640-63-9
:
:
C₇H₆Cl₂O₂S
:
MFCD03789183
:
225.09 g/mol
:
87 °C
:
GHS07
:
Ambient