3-Chloro-1H-indazole 95+%
Catalog # APOSOR7740-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:152.58 g/mol
Melting Pt:148…150 °C
Storage Temperature:Ambient
CAS Number:29110-74-5
Formula:C₇H₅ClN₂
MDL Number:MFCD00067527