Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Benzyl-1H-pyrazole-4-boronic acid
1-Benzyl-1H-pyrazole-4-boronic acid
1-Benzyl-1H-pyrazole-4-boronic acid
:
Apollo Scientific
1-Benzyl-1H-pyrazole-4-boronic acid
:
Apollo Scientific
:
852362-22-2
:
:
C₁₀H₁₁BN₂O₂
:
MFCD04038760
:
202.02 g/mol
:
76…81 °C
:
GHS07
:
Refrigerator