Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methoxy-3-pyridineboronic acid
4-Methoxy-3-pyridineboronic acid
4-Methoxy-3-pyridineboronic acid
Irritant
4-Methoxy-3-pyridineboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Methoxy-3-pyridineboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
355004-67-0
:
Formula:
C₆H₈BNO₃
MDL Number:
MFCD01114676
:
152.9436 g/mol
:
GHS07
Storage Temperature:
Freezer