Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Palmitoyl ceramide
N-Palmitoyl ceramide
N-Palmitoyl ceramide
N-Palmitoyl ceramide
Leverandør:
Apollo Scientific
Sertifikat
Utvid bildet
N-Palmitoyl ceramide
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
24696-26-2
:
Formel:
C₃₄H₆₇NO₃
MDL-nummer:
MFCD00871748
:
537.914 g/mol
Molten Pt:
94…95 °C
Lagringstemperatur:
Freezer