Order Entry
Norway
ContactUsLinkComponent
4-Pyrimidin-2-ylbenzoic acid 95%
Irritant
4-Pyrimidin-2-ylbenzoic acid 95%
Catalog # APOSOR6232-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Pyrimidin-2-ylbenzoic acid 95%
Catalog # APOSOR6232-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :200.1900 g/mol
Melting Pt:238…240 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:199678-12-1
Formula:C₁₁H₈N₂O₂
MDL Number:MFCD01318668