Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Dimethylamino-1-pyridin-2-ylpropenone
3-Dimethylamino-1-pyridin-2-ylpropenone 98%
3-Dimethylamino-1-pyridin-2-ylpropenone 98%
Irritant
3-Dimethylamino-1-pyridin-2-ylpropenone 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Dimethylamino-1-pyridin-2-ylpropenone 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
66521-54-8
:
Formula:
C₁₀H₁₂N₂O
MDL Number:
MFCD00115039
:
176.22 g/mol
Melting Pt:
126…130 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation